Sikkerhedsbriller, uvex overspecs
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9161.014 9161.305
111-8050EA 116 DKK
111-8050 111-1651
Sikkerhedsbriller, uvex overspecs
Briller
The cover glasses (OTG) are suited for a range of applications and fit over most prescription spectacles. Whether as visitor spectacles or as overspecs for short-term tasks, they allow perfect vision due to the large panoramic lenses.
  • Unrestricted peripheral vision
  • Pressure-free fit through duo-flex side arms
  • Side arm length adjustment
  • Integrated brow guard
Weight: 39 g
EN 166
Certificeringer: EN 166