Beskyttelsesbriller, uvex i-vo
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9160.065 9160.076 9160.265 9160.275 9160.520 9160.285 9160.120
111-8011EA 259 DKK
111-8011 111-8010 111-0144 111-1202 111-1215 111-1574 111-2300
Beskyttelsesbriller, uvex i-vo
Briller
These modern safety glasses offer exceptional comfort through duo-component technology - the combination of two materials (hard and soft). They combine optimum protection and fit with maximum comfort for the wearer.
  • High level of protection from mechanical and optical hazards, due to high impact resistant polycarbonate lenses
  • Indirect ventilation for maximum freedom from misting and increased safety
  • Angle and length of side pieces are individually adjustable for a comfortable and secure fit
Weight: 51 g
EN 166-168, EN 170; (EN 172: Anti-UV 5-2,5); 1F
Certificeringer: EN 166, EN 170; (EN 172: Anti-UV 5-2.5); 1F