Spoon spatulas
Leverandør: HAMMACHER KARL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HWN106-15 HWN107-18 HWN108-21
231-0263EA 488 DKK
231-0263 231-0264 231-0265
Spoon spatulas
Spatler
Double ended, broad spoon spatulas made of WIRONIT steel (stainless steel 18/12 with 18% chrome and 10 to 12% nickel).
  • Very resistant to corrosion and anti-magnetic
  • With one flat spatula end and one spoon end