Spoon spatulas, micro
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
985.130 985.150 985.185 985.210
231-2151EA 34.6 DKK
231-2151 RSGA985.150 231-2152 231-2153
Spoon spatulas, micro
Spatler
Stainless steel 18/10, anti-magnetic.
  • With round handle and one bent spoon
  • Excellent temperature resistance, autoclavable
  • Good resistance to chemicals
  • Very flexible