Double spatulas, micro
Leverandør: RSG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
373.150 374.150 376.130 376.150 376.185 376.210 374.130 373.210 377.150 374.185 375.150
231-0425EA 280 DKK
231-0425 231-1412 231-2321 231-2322 231-2323 231-2324 231-0427 231-0426 231-0430 231-0428 231-0429
Double spatulas, micro
Spatler
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance