Microarray slides, Gene Frame® II
Leverandør: Thermo Fisher Scientific
Ratings:
Gennemsntilig vurdering: 3.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 0 Nej: 0 
biologie moléculaire / chef de projet / toulouse
Ja, jeg anbefaler
un peu difficile de placer la lamelle sans faire de bulles ou évacuer du liquide à l'extérieur mais parfaitement étanche.
AB-0576 AB-0577 AB-0578
732-4824EA 1180 DKK
732-4824 732-4825 732-4826
Microarray slides, Gene Frame® II
Objektglas Objektglas, Microarray
Microarray slides for in situ hybridisation and amplification with standard microscope slides.
  • Gas tight sealing prevents reagent loss due to evaporation
  • Withstand temperatures of up to 97 °C
  • Frame and coverslip can be removed simultaneously leaving no residue
  • Can be autoclaved or sterilised by UV irradiation
Leveringsinformation: All packs are supplied with 100 coverslips.