Weighing scoops
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05568006 05568010 05568003
611-9310EA 210 DKK
611-9310 611-9322 611-9309
Weighing scoops
Øser
Borosilicate glass.
  • Ideal for weighing small samples
  • The tubular stem of the scoop also works as a weighing funnel and facilitates emptying of the scoop