Sampling scoops, Sterileware®, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H369020000 H369040000 H369060000
231-0008EA 2800 DKK
231-0008 231-0009 231-0010
Sampling scoops, Sterileware®, SP Bel-Art
Øser
PS, hvid. Designet specielt til prøvetagning fra beholdere med bred åbning.
  • One-piece construction
  • For powder, granulates and liquids
  • Made from FDA grade materials
  • Small sizes (60 and 125 ml) have a long handle that increases the distance between the hand and sample
  • When placed on a flat surface they remain upright; can be used as weighing boats
  • Individually sterilised and packed in a polyethylene bag
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904