Scoops
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8800 8801 8802 8803 8804
231-0244EA 198 DKK
231-0244 231-0109 231-0245 231-0246 231-0206
Scoops
Øser
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling, anti-magnetic