Scoops
Leverandør: SCHROEDER WILESCO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4279135 4279185 4279210 4279250 4279310 4279350 4279400 4279430 4279110 4279155_U
231-0301EA 53.8 DKK
231-0301 231-0302 231-0303 231-0304 231-0306 231-0307 231-0308 231-0309 231-0310 231-0311
Scoops
Øser
Aluminium, smooth. For collecting or measuring powders, granulates or pastes.
  • Good chemical resistance
  • Excellent thermal resistance
  • Light, robust and easy to use