Scalpel blades
Leverandør: BAYHA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FIGUR11 FIGUR15 FIGUR18 FIGUR19 FIGUR20 FIGUR21 FIGUR22 FIGUR23 FIGUR24 FIGUR26 FIGUR27 FIGUR28 FIGUR29
233-5211EA 96.2 DKK
233-5211 233-5215 233-5218 233-5219 233-5220 233-5221 233-5222 233-5223 233-5224 233-5226 233-5227 233-5228 233-5229
Scalpel blades
Skalpeller Skalpelblade
Kulstofstål.
  • Individual sterile packaging
  • Easy and quick blade replacement
  • Suitable for handles 233-5201, 233-5202 and 233-5203