Stand bases
Leverandør: R L ENTERPRISES
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3107/1 3107/2 3107/21 3107/3 3107/31 3107/4
241-0173EA 552 DKK
241-0173 241-0176 241-0177 241-0179 241-7869 241-0182
Stand bases
Retortstativer
Rectangular retort bases with M10 threaded hole in centre of short side.
  • Steel, with acrylic finish
  • Acid resistant surface
  • Non-slip rubber feet for a firm footing
Bestillingsoplysninger: Supplied without rods which need to be ordered separately.