Tripod stands
Leverandør: BOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5251 5250
241-7123EA 617 DKK
241-7123 241-1109
Tripod stands
Retortstativer
Cast iron, varnished.
  • With M10 threaded hole
  • No support rod
  • Slip-proof, rubber feet ensure firm footing