Reaction vessels, cylindrical, with flat flange and thermal jacket
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06010137 06010149 06010258 06010270 06010273 06010473 06010479 06010485 06010482 06010158 06010249 06010276 06010279 06010476 06011494 06011588
LENZ06010137EA 4030 DKK
LENZ06010137 LENZ06010149 LENZ06010258 201-1157 201-1158 LENZ06010473 LENZ06010479 LENZ06010485 LENZ06010482 LENZ06010158 LENZ06010249 LENZ06010276 LENZ06010279 211-2050 LENZ06011494 LENZ06011588
Reaction vessels, cylindrical, with flat flange and thermal jacket
Reaktionsbeholdere Reaktionsbeholdere, cylinderformede
Reaction vessel, made of DURAN® borosilicate glass, flat bottom, with easily readable graduation and flat laboratory flange (DN) with groove. With two connectors with GL 18 thread.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes