Reaction vessels, cylindrical, with flat flange, thermal jacket and drain valve
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06030488 06031488 06031494 06031588 06031594 06030137-V2 06030149-V2 06030158-V2 06030249-V2 06030258-V2 06030270-V2 06030273-V2 06030276-V2 06030279-V2 06030473-V2 06030476-V2 06030479-V2 06030482-V2 06030485-V2 06031137-V2 06031149-V2 06031158-V2 06031249-V2 06031258-V2 06031270-V2 06031273-V2 06031276-V2 06031279-V2 06031473-V2 06031476-V2 06031479-V2 06031482-V2 06031485-V2 06031585-V2
LENZ06030488EA 23370 DKK
LENZ06030488 LENZ06031488 LENZ06031494 LENZ06031588 LENZ06031594 LENZ06030137-V2 LENZ06030149-V2 LENZ06030158-V2 LENZ06030249-V2 LENZ06030258-V2 LENZ06030270-V2 LENZ06030273-V2 LENZ06030276-V2 LENZ06030279-V2 LENZ06030473-V2 LENZ06030476-V2 LENZ06030479-V2 LENZ06030482-V2 LENZ06030485-V2 LENZ06031137-V2 LENZ06031149-V2 LENZ06031158-V2 LENZ06031249-V2 LENZ06031258-V2 LENZ06031270-V2 LENZ06031273-V2 LENZ06031276-V2 LENZ06031279-V2 LENZ06031473-V2 LENZ06031476-V2 LENZ06031479-V2 LENZ06031482-V2 LENZ06031485-V2 LENZ06031585-V2
Reaction vessels, cylindrical, with flat flange, thermal jacket and drain valve
Reaktionsbeholdere Reaktionsbeholdere, cylinderformede
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With easily readable graduation and flat laboratory flange (LF) with groove. With two connectors with GL 14/GL 18 or KF 15/KF 25 ball flanges.