Reaction vessels, cylindrical, with drain valve
Leverandør: Lenz Laborglas GmbH & CO.KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06007488 06007494 06007588 06007594 06007128-V2 06007137-V2 06007149-V2 06007158-V2 06007258-V2 06007270-V2 06007273-V2 06007276-V2 06007279-V2 06007473-V2 06007476-V2 06007479-V2 06007482-V2 06007485-V2 06007585-V2
LENZ06007488EA 16880 DKK
LENZ06007488 LENZ06007494 LENZ06007588 LENZ06007594 LENZ06007128-V2 LENZ06007137-V2 LENZ06007149-V2 LENZ06007158-V2 LENZ06007258-V2 LENZ06007270-V2 LENZ06007273-V2 LENZ06007276-V2 LENZ06007279-V2 LENZ06007473-V2 LENZ06007476-V2 LENZ06007479-V2 LENZ06007482-V2 LENZ06007485-V2 LENZ06007585-V2
Reaction vessels, cylindrical, with drain valve
Reaktionsbeholdere Reaktionsbeholdere, cylinderformede
Reaction vessel, made of DURAN® borosilicate glass, round bottom, with easily readable graduation and laboratory flat flange (LF) with groove.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes