Flat flange lids with 5 necks, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MAF3/52 MAF5/2_U MAF2/2 MAF1/75 MAF2/52 MAF2/41
201-8582EA 2100 DKK
201-8582 212-0615 201-8578 201-8577 201-8581 BIBBMAF2/41
Flat flange lids with 5 necks, Quickfit®
Låg Reaktionsbeholderlåg
Lids, made of PYREX®, Borosilicate glass, with precision ground, flat flange joints.
  • For use with Quickfit® reaction flasks and culture vessels
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
* 201-8578 has 2× side (5°) sockets should be secured to flasks with relevant metal clips 201-8565 or 201-8562.