Flat flange lids with 1 neck, Quickfit®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DA4/100 DA5/100 DA5/75
201-0257EA 1580 DKK
201-0257 201-0525 201-0258
Flat flange lids with 1 neck, Quickfit®
Låg Reaktionsbeholderlåg
PYREX®, borosilicate glass.
  • For use with Quickfit® wide neck reaction flasks and culture vessels
  • Excellent chemical resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With precision ground, flat flange joint.