Test Tube racks, wire
Leverandør: VICTORSSON INDUSTRIER
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4X6X13X35 4X15X13X35 3X6X13X50 4X6X20X75 2X10X20X75 3X10X20X75 4X12X20X75 4X6X32X100 2X6X43X75 3X5X17X45 4X11X17X45 2X6X30X80 4X15X13X50 3X4X20X75 3X6X20X75 2X10X20X60
525-0374EA 1000 DKK
525-0374 525-0372 525-0366 525-0376 525-0354 525-0360 525-0371 525-0378 525-0358 525-0365 525-0369 525-0357 525-0373 525-0364 525-0368 525-0353
Test Tube racks, wire
Stativer Tube Racks
Nylon coated wire, autoclavable. Longitudinal wires across base.