Racks for test tubes
Leverandør: HUPFER METALLWERKE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7500634 7500784 7500631 7500647 7500633 7500632 7500649 7500785 7500786 7500651 7500650 7500628 7500627 7500648
212-6405EA 606 DKK
212-6405 212-6410 212-6415 212-6420 212-6425 212-6430 212-6455 212-6460 212-6461 212-6465 212-6470 212-6475 212-6480 212-6489
Racks for test tubes
Stativer Tube Racks
Stainless steel, rust-proof, electrolytically or steel polished, polyamide coated.
  • Excellent chemical resistance
  • Excellent thermal resistance (polyamide coating resistant up to 80 °C)
  • Robust, stable construction and easy to clean
Trådracks med to etager, højde 21 mm. Samlet højde 68 mm. Hulstørrelse 18×18 mm.