Racks til reagensglas, radioimmunassay, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F188720000 F188710000 F188700000
212-6501EA 2240 DKK
212-6501 212-6502 212-6503
Racks til reagensglas, radioimmunassay, SP Bel-Art
Stativer Tube Racks
PP, hvid.
  • Temperaturresistent op til 121 °C, kan steriliseres med damp
  • God kemisk resistens
These racks provide a practical method of pipetting and storing a large number of samples. By using alternate rows and rotating the tubes, it is easy to keep track of the last tube filled. Also useful in clinical laboratories for storage of samples.