Microtube and test tube racks, round, SP Bel-Art
Leverandør: Bel-Art Products, a Part of SP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F187431016 F187431020 F187410020
212-9260EA 468 DKK
212-9260 212-6264 212-9243
Microtube and test tube racks, round, SP Bel-Art
Stativer Tube Racks
PP, red or white.
  • Meget god kemisk resistens
  • Varmeresistent op til 120 °C, autoklaverbar
  • Meget stabil, brudsikker
Versatile. Practical for applications where you experience problems when using square racks. Ideal for use in water baths. Alphanumeric position designators for easy sample identification. Stackable.