VWR®, Racks til reagensglas
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0192EA 873 DKK
525-0192 525-0193 525-0194 525-0195
VWR®, Racks til reagensglas
Stativer Tube Racks
PTFE.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Non stick surfaces for easy cleaning
Robust and inert to virtually all chemical reagents, with very good thermal resistance. Suitable for use in cryogenics, freezers and ovens.