Racks for microcentrifuge tubes, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RPB001P RPW001P
AZLORPB001PEA 1280 DKK
AZLORPB001P AZLORPW001P
Racks for microcentrifuge tubes, Azlon®
Stativer Tube Racks
PP, autoklaverbare.
  • Designed for stable use in water baths
  • Stackable when empty or full
  • Alphanumeric grid reference aids tube identification