Vial racks, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
868804 868808 868810 985750
548-2157EA 3290 DKK
548-2157 548-1976 548-2158 548-2156
Vial racks, WHEATON®
Stativer Stativer til Kromatografirør
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
  • Autoclavable and stackable
  • Good chemical resistance
  • Includes position identifiers