VWR®, Racks for Centrifuge Tubes and Bottles
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
525-0741EA 261 DKK
525-0741 211-0197 525-0742
VWR®, Racks for Centrifuge Tubes and Bottles
Stativer Tube Racks
Epoxybelagt stålwire.
  • Ribbet bund for bedre stabilitet af rørene
  • Meget god kemisk resistens
Racks for 15 and 50 ml centrifuge tubes and 250 ml centrifuge bottles. Can also be used for all other types of tubes.