TLC-plader, aluminiumoxid 60
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.05550.0001 1.05581.0001 1.05713.0001 1.05731.0001
1.05550.0001EA 2390 DKK
1.05550.0001 1.05581.0001 1.05713.0001 1.05731.0001
TLC-plader, aluminiumoxid 60
TLC-plader
Merck TLC aluminium oxide plates use neutral or basic aluminium oxide of 60 or 150 Å pore size with or without fluorescent indicator to suit different application needs. Under aqueous conditions, basic compounds can be best separated on basic aluminium oxide plates, while neutral compounds are best separated on neutral plates.
  • Tydelig separationsfordele med hensyn til pH