Robotic tips for PerkinElmer®/Packard® (PlateTrak/MiniTrak and Evolution)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
925-021 925-261 925-262 927-021 927-261 927-262 929-021 929-261 929-262
736-0025EA 2070 DKK
736-0025 737-2003 737-2004 736-0026 737-2005 737-2006 736-0027 737-2007 737-2008
Robotic tips for PerkinElmer®/Packard® (PlateTrak/MiniTrak and Evolution)
Pipettespidser
High quality tips, intended for research in molecular biology laboratories.
  • Transparent PP
  • Certified free of RNase, DNase, DNA and pyrogens
  • Very low coefficient of variation
  • Tips with ART® filters: Eliminate any risk of cross-contamination between samples; designed for applications with DNA, RNA as well as radioactive and infectious samples
  • Sterile tips: Sterilised by gamma radiation