Pipettespidser, epT.I.P.S.® Standard
Leverandør: EPPENDORF
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0030000.870 0030000.897 0030000.919
613-3503EA 330 DKK
613-3503 613-3504 613-3505
Pipettespidser, epT.I.P.S.® Standard
Pipettespidser
These tips feature an ergonomically optimised cone geometry which provides an improved tip seal.
  • Kvalitetsspidser
  • Ergonomisk design
  • Kegleform med smidig og bøjelig forseglingsområde, som giver en bedre forsegling ved pipetteskaftet
epT.I.P.S., in combination with adequate pipettes, are intended for dispensing liquids in the volume range of 0,1 µl to 10 ml.
In vivo applications (applications in or on the human body) are not allowed. epT.I.P.S. and the appropriate pipettes only must be used by correspondingly trained specialists.
The user mustMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable Lav fremstillingspåvirkning
Miljøcertificering Renewable Energy Certificate, ISO 14001