Skyllesæt til pipetter
Leverandør: KARTELL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215 217 218 219 221 222
149-1111EA 2940 DKK
149-1111 149-1110 149-1122 149-1131 149-1121 149-1130
Skyllesæt til pipetter
Pipetter Pipettevasker
Rinsing sets for cleaning pipettes up to 600 mm length and Mohr burettes gently and efficiently. The entire washing system consists of a washer, a pipette jar and a pipette basket.
  • Heat resistant up to +105 °C
  • Good chemical resistance
  • Emptying siphon allows constant water exchange
  • Soft PE base of the pipette basket minimises breakage
  • Basket is provided with a height-adjustable ring to hold the pipettes together during the removal from the pipette washer
Bestillingsoplysninger: All components need to be ordered separately.