Graduated pipettes, type 3, Class B, MBL
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PRA830B PRA832B PRA834B PRA838B PRA836B
612-2945EA 168 DKK
612-2945 612-2946 612-2947 612-2949 612-2948
Graduated pipettes, type 3, Class B, MBL
Pipetter Pipetter med inddeling
Soda lime glass.
  • Calibrated to deliver from any graduation line down to zero at the jet
  • Graduations in permanent amber stain
BS700, ISO 835
Certificeringer: BS700, ISO 835