Bulb pipettes, one mark, class AS, blue graduation
Leverandør: HIRSCHMANN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1341133 1341140 1341145 1341148 1341155 1341160 1341164 1341165 1341170 1341175 1341180
612-1310EA 69 DKK
612-1310 612-1311 612-1312 612-1313 612-1314 612-1315 612-1316 612-1317 612-1318 612-1384 612-1320
Bulb pipettes, one mark, class AS, blue graduation
Pipetter Kolbepipetter
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
DIN EN ISO 648
Certificeringer: DIN EN ISO 648; DE-M marking