Fuldpipetter, BLAUBRAND®, 2 mærker, klasse AS
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
29721 29722 29723 29725 29727 29732 29733 29734 29735 29738
612-2726EA 89.3 DKK
612-2726 612-2727 612-2728 612-2729 612-2730 612-2731 612-4759 612-2732 612-2733 612-2734
Fuldpipetter, BLAUBRAND®, 2 mærker, klasse AS
Pipetter Kolbepipetter
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in high contrast blue enamel
  • Including one batch certificate
DIN EN ISO 648
Certificeringer: DIN EN ISO 648; DE-M marking.