Fuldpipetter, BLAUBRAND®, 1 mærke, klasse AS
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
29701 29702 29703 29704 29705 29706 29707 29708 29709 29710 29711 29712 29713 29714 29715 29716 29717 29718 29719
612-1201EA 74.9 DKK
612-1201 612-1202 612-1203 612-1204 612-1205 612-1206 612-1207 612-1208 612-1209 612-1210 612-1211 612-1212 612-1213 612-1214 612-1215 612-1216 612-1217 612-1218 612-1219
Fuldpipetter, BLAUBRAND®, 1 mærke, klasse AS
Pipetter Kolbepipetter
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time: 5 seconds
  • Batch certificate
  • Also available (on request) with an individual certificate or DAkkS calibration certificate
  • USP version available
DIN EN ISO 648
Certificeringer: DIN EN ISO 648; DE-M marking.