Dækglas
Leverandør: MARIENFELD
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0101000 0101062 0101092 0101122 0101142 0101152 0101182 0101202 0101212 0101020 0101050 0101010 0101030 0101040 0101060 0103990091
630-1585EA 254 DKK
630-1585 630-1588 630-1589 630-1590 630-1591 630-1592 630-1593 630-1594 630-1595 630-1586 631-0653 631-0655 631-0656 631-0657 631-0659 MARI0103990091
Dækglas
Dækglas
Made of chemically resistant borosilicate D 263™ M glass, Hydrolytic Class I
  • No bubbles or flaws
  • Suitable for in vitro diagnostic (IVD) applications
Thickness: 0,13 - 0,16 mm
Emballering: Pakket i æske med separat låg.