96-brønds PCR-racks
Leverandør: HEATHROW
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
HS23461C HS23461D HS23461B HS23461A
211-3501EA 59 DKK
211-3501 211-3502 211-3504 211-3505
96-brønds PCR-racks
Stativer Tube Racks
High resistance PP.
  • Tube wells are easily identified by imprinted numbers and letters
  • Removable, hinged lid
Designed to accommodate 96 individual 0,2 ml tubes or 12 or 8 strips of 8 or 12 tubes.
Colour: Natural, blue, green, yellow or assorted colours (one each of blue, green, pink, yellow and natural).