PCR-plader, 24 og 48 brønde, Thermo-Fast®
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AB-0624 AB-0648
732-4837EA 1240 DKK
732-4837 732-4841
PCR-plader, 24 og 48 brønde, Thermo-Fast®
Mikroplader PCR plader
PP. Egnede til 0,2 ml thermocyclerblokke.
  • Sealable using adhesive films and foils, heat seals and flat or domed cap strips
  • Maximum well volume 0,3 ml when used with adhesive and heat seals
  • Certified free from DNase, RNase and human genomic DNA
Colour: Natural