96-Multiwell insert system, Falcon®, Corning
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Falcon®
734-0368EA 18340 DKK
734-0368 734-1379
96-Multiwell insert system, Falcon®, Corning
Mikroplader Indsatsplader
The Falcon® HTS 96-multiwell insert system is a cell culture insert platform suitable for both manual and robotic screening of compounds in cell-based assays. The system has been tested for its ability to produce a differentiated monolayer of Caco-2, LLC-PK1, and MDCK cells making it an ideal platform for in vitro bioavailability and permeability studies. This automation compatible platform is composed of a 1,0 µm pore size PET membrane-based 96-multiwell insert plate, a media feeder tray and a lid. The drop-in baffle for the feeder tray mitigates media sloshing and lowers the risk of contamination.
  • Automation compatible design suitable for use with most robots and fluid handling instruments
  • Excellent reproducibility - the one-piece feeder tray enhances consistency in well-to-well monolayer growth
  • Ideal for drug transport studies