Cell culture plates, 1536-well, CELLSTAR®
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
782093_60 782092_60 783092_60 782073_60 782078_60 782080_40 782086_40
738-0075EA 38620 DKK
738-0075 738-0074 738-0085 738-0066 738-0071 738-0072 738-0073
Cell culture plates, 1536-well, CELLSTAR®
Mikroplader Celledyrkningsplader
PS, cell culture-treated, sterile. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • Physically surface treated for improved cell adhesion
  • HiBase versions designed for top reading instruments, µClear® LoBase plates for bottom reading instruments
  • Rounded square well design combines the advantages of the square well (flexible working volume, 3 to 10 µl) with the advantages of a round well (reduced wicking and bubbling)
Colour: μClear® (clear), black or white