Cell culture plates, 96-well, CELLSTAR®
Leverandør: Greiner Bio-One
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
655180_100 655162_100 655182_160 655185_60 651180_100 650160_100 650180_100 651160_100 655160_100 650185_240 655180_NOV
391-3325EA 2190 DKK
391-3325 392-0283 392-0225 392-0284 392-0021 392-0018 392-0019 392-0020 392-0026 GREI650185_240 GREI655180_NOV
Cell culture plates, 96-well, CELLSTAR®
Mikroplader Celledyrkningsplader
PS, cell culture-treated, sterile. Cell culture treated plates are available with round (U), conical (V) or flat (F) bottom wells. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • U-bottom plates have no sharp corners, so can be pipetted easily and cleanly
  • V-bottom plates can be pipetted precisely
  • F-bottom plates have excellent optical properties, as well as a larger working volume
  • The chimney well design of the F-bottom plate prevents cross contamination
  • Lid enables gas exchange with minimal evaporation
Colour: Clear
With condensation rings.