Cellekulturinserts til Companion-plader, Falcon®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
353502 353503 353504 355467
734-0065EA 1470 DKK
734-0065 734-0066 734-0067 734-1095
Cellekulturinserts til Companion-plader, Falcon®
Mikroplader Indsatsplader
PS, tissue culture-treated, sterile, non-pyrogenic, with lid. Specifically designed for use with Falcon® or Corning® BioCoat™ cell culture inserts, so that evaporation and contamination due to improper lid fit is eliminated.
  • Reagents can be added quickly and consistently for timed experiments
  • Aspiration from the well is easier, reducing the risk of contamination
  • Media cannot wick up between the insert and the well wall
  • Low-evaporation lid reduces evaporation and contamination
In the 'Feeding Position' pipette access is improved for fluid handling on the basolateral side. In the 'Incubation Position' cell culture inserts remain locked in position in their companion plate wells.
Note: May be used with or without cell culture inserts.