Microtitre plates, 96-well
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2605 2205 9205
735-0184EA 761 DKK
735-0184 735-0180 735-0194
Microtitre plates, 96-well
Mikroplader HTS mikroplader
Rigid styrene or flexible vinyl, square well. Ideal for micro-volume applications, such as storage, sample or reagent transfer, serial dilution, haemogglutination, blood typing and compliment fixation. Available with a choice of flat, U- or V-bottom well shape.
  • U-bottom well shape improves washing in ELISAs, enhances sensitivity in fluorescent applications and facilitates observation of agglutination reactions
  • V-bottom well shape is optimal for magnetic bead assays, ideal for agglutination tests, useful for centrifugation or concentration and facilitates recovery of contents