Measuring cylinders, tall form, class B, PYREX®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3205/04MP 3205/06MP 3205/08P 3205/10P 3205/12P
612-0065EA 170 DKK
612-0065 612-0067 612-0069 612-0071 612-0073
Measuring cylinders, tall form, class B, PYREX®
Cylindere Målecylindre
Borosilicate glass, tall form, class B.
  • White enamel graduations
  • Polyethylene foot absorbs impact and reduces breakage