Measuring cylinders, tall form, class B, borosilicate glass, heavy duty, PYREX®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3280-50 3280-100 3280-250 3280-500 3280-1L 3280-2L
612-0179EA 705 DKK
612-0179 612-0172 612-0177 612-0204 612-0174 BIBB3280-2L
Measuring cylinders, tall form, class B, borosilicate glass, heavy duty, PYREX®
Cylindere Målecylindre
Borosilicate glass, tall form, with spout, class B.
  • Thick wall
  • Robust hexagonal foot
  • Graduations in high contrast blue enamel
ISO 4788
Certificeringer: ISO 4788