Measuring cylinders, tall form, class B, PP, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CP0050P CP0100P CP0250P CP0500P CP2000P CPB0010P CPB0025P CPB0050P CPB0100P CPB0250P CPB0500P CPB1000P CPB2000P CP0025P CP1000P CP0010P
612-0379EA 524 DKK
612-0379 612-0380 612-0387 612-0388 612-0390 612-0399 612-0402 612-0403 612-0404 612-0405 612-0406 612-0407 612-0408 612-0378 612-0389 612-0377
Measuring cylinders, tall form, class B, PP, Azlon®
Cylindere Målecylindre
PP, highly translucent, tall form, class B.
  • Hexagonal base with non-slip, anti-suction feature, spout, extremely durable
  • Recycling codes and batch code on each cylinder
  • Autoclavable, but accuracy may be affected
ISO 6706
Certificeringer: ISO 6706