Measuring cylinder, tall form, class B, PP
Leverandør: VITLAB
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
646941 647941 648941 649941 650941 651941 652941 653941
612-2426EA 27.3 DKK
612-2426 612-2427 612-2428 612-2429 612-4354 612-4355 612-4356 612-4357
Measuring cylinder, tall form, class B, PP
Cylindere Målecylindre
PP, gennemskinnelig, høj form, klasse B.
  • Calibrated to 'In'
  • Hexagonal base, ring mark on the principal points
  • Thermal stresses of up to +80 °C do not result in the tolerance limit being permanently exceeded
Certificeringer: DIN 12681 B / ISO 6706