Beholder, bred hals, UN-godkendt, med skruelåg
Leverandør: CURTEC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
7006-61-046 7010-61-046 7020-61-046 7003-61-046 7015-61-046 7026-61-046 7042-91-046 7055-91-046 7068-91-046 7011-61-046
216-5802EA 357 DKK
216-5802 216-5803 216-5806 216-5801 216-5804 216-5805 216-4225 216-4226 216-4233 216-2055
Beholder, bred hals, UN-godkendt, med skruelåg
Tønder, fustager, trommer
HDPE drums, with screw lid.
  • Water tight
  • Tamper-evident
  • Stackable
  • UN marking (1H2) for hazardous solids
  • Food grade
For storage and shipment of powders, pastes and liquids.