VWR® Safety-Blue™, Thermo-Hydrometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-0924EA 799 DKK
620-0924 620-0925 620-0926 620-0928 620-0929
VWR® Safety-Blue™, Thermo-Hydrometers
Flydevægte
These safety sugar scale combined form thermo-hydrometers can determine the specific gravity, Brix and alcohol by volume.
  • Lead free ballast and glass with mercury-free blue spirit organic liquid fill
  • Individually serialised, safe and easy to clean up in the event of a breakage
  • Temperature standardisation of 20 °C with thermometer
Designed to measure sucrose in the food and beverage and other industries.
Certificeringer: ASTM
Leveringsinformation: Includes a multi-language statement of accuracy indicating traceability to NIST.