VWR®, Aræometre, M50
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
614-0311EA 224 DKK
614-0311 614-0312 614-0313 614-0314 614-0315 614-0316 614-0317 614-0318
VWR®, Aræometre, M50
Flydevægte
Hydrometers with specific gravity.
  • Conform to ISO 649 (BS 718, DIN 12791, NF B 35511)
  • Calibration temperature: 20 °C
  • Division 0.001 g/ml
Length: 270 mm
Certificeringer: ISO 649. BS 718, DIN 12791, NF B 35511.
Emballering: In a strong transparent plastic case.