Engangs-baseforme
Leverandør: Simport Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M475-1 M475-3 M475-10 M475-4 M475-5 M475-2
720-0820EA 3620 DKK
720-0820 720-0822 720-0208 720-0823 720-0824 720-0821
Engangs-baseforme
Forme Forme til histologi
These disposable PVC base moulds offer ease and convenience of operation. They can be discarded after use, but are also strong enough to be reused.
  • Excellent thermal exchange
  • Smooth interior
  • Rounded corners
They are available in the same variety of sizes as metal moulds and can be used with the same styles or types of cassettes and embedding rings.
Leveringsinformation: Each case contains two dispenser boxes of 500 base moulds.